Eisco Metal Support Rods with Threaded Ends
Supplier: Wards
Certificates
About this item
Heavy metal rods with threaded ends that screw into retort bases.
- Available in high quality aluminum, steel, or stainless steel
- Measures 12 mm in diameter and come in a range of weights
- Threaded end (10×1.5 mm) fits in all Eisco retort bases
Support provide superb stability for laboratory experiments and setups.
Customers Who Bought This Also Bought