Sepharose™ Streptavidin High Performance Affinity Chromatography Media
Catalog #: CA76488-174
Supplier: Cytiva
CAS Number:
Specifications
- Packaging:25 ml
- Particle size:34 µm
- pH range:4 - 9
- Description:Streptavidin Sepharose™ High Performance
- Flow rate:<150 cm/h
- Matrix:Highly cross-linked agarose, 6%
- Storage temperature:4...8 °C
- Cat. no.:CA76488-174
- Type:Liquid
Specifications
About this item
Streptavidin Sepharose™ High Performance is an affinity chromatography resin designed for fast and reliable, high resolution purification of biotinylated biomolecules.
Extremely useful for exploiting eith.er the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin
34 µm bead size for improved resolution.
Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose™ High Performance. The base matrix is a rigid, highly cross-linked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.
Customers Who Bought This Also Bought