Drive Sleeves
Catalog #: CA10498-032
Supplier: Cytiva
Specifications
- Description:Drive sleeve, without wetted part
- Cat. no.:CA10498-032
- Supplier No.:19-6067-02
Specifications
About this item
Customers Who Bought This Also Bought
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
$81.84 to $877.92
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$214.10 to $1,039.32
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Masterflex® Ismatec® Replacement Cartridges and Adapters, Avantor®
Supplier: Avantor Fluid Handling
$139.04
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$35.22 to $841.56
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
$41.13 to $265.86