To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:5.000 EU
- Enzyme name:Exonuclease I
- Cat. no.:CA95040-316L
- Supplier No.:E70073X
Specifications
About this item
Exonuclease I acts specifically on single-stranded DNA degrading it processively in the 3'- to 5'-direction, producing 5'-mononucleotides. Applications include eliminating residual single-stranded DNA containing a 3'-terminus, measuring endonucleolytic cleavage of covalently closed circular (ccc) ssDNA, measuring DNA helicase activity.
- 3'→5' single-stranded exonuclease
- Inactivated by heating at 80 °C for 15 minutes
- Active in PCR buffers
- Easily soluble in cold water and hot water
Digestion of single-stranded DNA by Exonuclease I releases deoxyribonucleoside 5'-mono-phosphates leaving 5'-terminal dinucleotides intact. DNA strands with terminal 3'-OH groups blocked by phosphoryl or acetyl groups are not removed. Monomeric 54.5 kDa bacterial exonuclease I recombinantly expressed in E. coli. One unit of the enzyme catalyzes the release of 10 nmol of acid soluble nucleotides in 30 minutes at 37 °C.
Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases. Greater than 95% pure as determined by SDS-PAGE.
illustra Exonuclease I, used together with illustra Alkaline Phosphatase for inactivation of dNTPs, provides effective cleanup of PCR reactions for subsequent use in DNA sequencing.