HyperSep™ Aminopropyl SPE Plates
Supplier: Thermo Scientific
Certificates
Certificates
About this item
A polar sorbent for both polar and anion exchange interactions.
- Particle size: 40 to 63 µm
- Pore size: 58 Å
- Surface area: 496 m²/g
Customers Who Bought This Also Bought
Hydrochloric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Supplier: VWR International
99,45 $ to 1 061,29 $
SensaGuard™ Polyethylene Gloves, Disposable, Industrial Grade Embossed, MCR Saftey
Supplier: MCR Safety
47,51 $ to 532,63 $
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
428,18 $
EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
Supplier: Honeywell Research Chemicals
153,96 $
Choline chloride 98.0-100.5% (by anhydrous basis), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
149,28 $
Media/Lab Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
103,73 $ to 117,47 $
Puritan® UniTranz-RT® Media Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
304,06 $ to 1 516,46 $