Specifications
- Description:GEM RUNNER™ dispenser
- Cat. no.:76717-342
Specifications
About this item
GEM RUNNER™ features a reloadable single edge blade safety dispenser for cleanroom and medical manufacturing applications.
- Holds all GEM3 and GEM2 cartridges
- Fits 0.009" and 0.012" single edge blade GEM cartridges
- Designed for safety and speed
- Great for high volume uses
- Blades not included
GEM RUNNER™ reduces the risk of injury, streamlines cutting operations, and minimizes waste. It serves up blades at the touch of a button. It’s compact and reliable, and it’s the only reloadable pop-up blade dispenser on the market.