About this item
Gain proficiency in micropipetting through three engaging activities.
Whether determining the distance to a faraway star or weighing the correct dose of a drug for a test subject, accurate and precise measurements are a critical part of any scientic investigation. In your kitchen, you might make accurate and precise measurements using tools like measuring cups and measuring spoons. When scientists need to accurately measure small volumes of liquid, they use a tool called a micropipette. Prepare for your future career in the lab and become a micropipette expert by completing the included activities.
Micropipetting practice: This is the first step on your way to becoming a micropipette expert. Practice precisely measuring and mixing small volumes using a pipette practice card.
Gel loading practice: Next, learn the art of loading an electrophoresis gel, a critical tool in the molecular biology lab. Instead of the agarose gel you might encounter in the lab, you’ll use a sturdy, reusable silicone practice gel.
Micropipette art: Finally, put your micropipetting skills to the test and employ your creativity to create micropipette masterpieces. Afterwards, share your artwork with us on Instagram for a chance to win a prize!
Storage: All materials can be stored at room temperature.