Specifications
- Description:Used Blade Container, plastic
- Cat. no.:61200-328
- L×W×H:107×105×210 mm
- Supplier No.:9810
Specifications
About this item
You can fill this container with the content of your Safebox. Or directly with your used blades.
When the box is full, turn the lid until it shuts. Now the box cannot be reopened. The next step is to safely and completely dispose of the filled Used Blade Container as normal waste.