TLC Plates
Supplier: Buchi
New Product
Certificates
About this item
Glass or aluminum backed plates coated with FlashPure ID Silica with fluorescent indicator, UV 254 nm.
- For normal phase applications
Quickly perform method development for flash chromatography separations. Monitor eluting fractions and determine which fractions contain desired materials.
Specifications
- Catalog No:
- 76836-782
- Particle size distribution:
- 6 - 10 µm