About this item
The Pure Essential Flash Chromatography System focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
- 50 bar (725 psi) pump, pressure sensors, and regulators
- 4 fixed UV wavelengths for most common applications
- Liquid or solid sample loading
- Cartridges from 4 to 5000 g or glass columns
- Enclosed and vented fraction collector bay
The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable. Basic but powerful and safe thanks to a 3-piston pump. Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900 pump.
Intuitive navigation is provided by novel and state-of-the-art software (methods, run data, and general settings). All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow.