Specifications
- Compatible with:All flow cells
- Description:LFB expansion
- Cat. no.:76848-064
- No. of Reactions:4
Specifications
About this item
Additional Long Fragment Buffer (LFB).
The LFB Expansion contains additional Long Fragment Buffer (LFB) for use with sequencing experiments where shorter fragments (<3 kb) are to be removed. The LFB Expansion contains enough reagents for four reactions.