Specifications
- Compatible with:All flow cells
- Description:Short fragment eliminator expansion
- Cat. no.:76848-058
- No. of Reactions:30
Specifications
About this item
An expansion pack designed for size selection of high molecular weight DNA samples to progressively deplete short fragments of <25 kb.
- Size select to progressively deplete short fragments (<25 kb) prior to sequencing library preparation
- Enrich sequencing libraries for long fragments without using any external third-party reagents
The Short Fragment Eliminator (EXP-SFE001) Expansion pack is recommended for users who want to size select high molecular-weight (HMW) gDNA to deplete short fragments (<25 kb) using size selective precipitation. The kit contains Short Fragment Eliminator (SFE) buffer which progressively removes fragments up to 25 kb, with fragments under 10 kb being almost completely removed in a quick and simple workflow.
Note: This kit is designed to remove short DNA fragments and requires samples to be predominantly made up of HMW DNA (>48 kb). If there are large amounts of short fragments in your sample, this will result in a low yield at the end of the prep and a low sequencing output.