Sepharose™ CL-4B Octyl Hydrophobic Interaction Chromatography Media, Cytiva
Catalog #: CA95016-994L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:90 µm
- pH range:3 - 12
- Description:Octyl Sepharose™ CL-4B
- Flow rate:150 cm/h
- Size:200 mL
- Matrix:Cross-linked agarose, 4%, spherical
- Ligand:Octyl
- Storage temperature:4 - 30 °C
- Ligand density:40 μmole/ml drained medium
- Description:Octyl Sepharose™ CL-4B
- Cat. no.:CA95016-994L
- Type:Liquid
- Supplier No.:17-0790-01
Specifications
About this item
Octyl-Sepharose™ CL-4B is a well-proven medium for hydrophobic interaction chromatography (HIC). Octyl immobilised on 4% cross-linked agarose.
- Phenyl immobilized on 4% cross-linked agarose
- Aromatic ligand
- Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
- Negligible volume variation due to changes in pH or ionic strength
Octyl Sepharose® CL-4B and Phenyl Sepharose® CL-4B are covalently coupled to a cross-linked 4% agarose matrix by ether linkage, resulting in hydrophobic gels with minimal leakage and no ionic properties.
Customers Who Bought This Also Bought