Sepharose™ CL-2B Gel Filtration Base Matrix
Catalog #: CA95056-222L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:60 - 200 μm
- pH range:3 - 13
- Description:Sepharose™ CL-2B
- Flow rate:15 cm/h
- Size:1 L
- Matrix:2% agarose
- Storage temperature:4 - 30 °C
- Description:Sepharose™ CL-2B
- Cat. no.:CA95056-222L
- Type:Liquid
- Exclusion Limit:>4 × 107
- Supplier No.:17-0140-01
Specifications
About this item
Sepharose CL-2B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
- 2% cross-linked agarose gel filtration media
- Proven base matrix for coupling affinity ligands
- Higher degree of crosslinking improve matrix stability
Customers Who Bought This Also Bought