illustra™ MicroSpin™ S-300 HR columns, Cytiva
Catalog #: CA95017-617L
Supplier: Cytiva
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:Illustra™ MicroSpin™ S-300 HR columns
- Size:50 Preps
- Sample size:25 to 50 µL
- Description:Illustra™ MicroSpin™ S-300 HR columns
- Cat. no.:CA95017-617L
- Supplier No.:27-5130-01
Specifications
About this item
illustra™ MicroSpin™ S-300 HR columns designed for rapid purification of PCR products (>100 bp) from unincorporated primers (<20-mers) and nucleotides using spin-column chromatography.
- Accommodates 25 to 50 μl for post-PCR clean-up prior to sequencing
- Useful for purification of alkaline-denatured plasmid DNA prior to sequencing
- Prepacked for convenience with Sephacryl™ S-300 HR pre-equilibrated in TE buffer
- Ready to use, requiring less than 4 minutes from sample application to collection of purified product
- Tested in nickase, single- and double-stranded exonuclease and RNase assays
Customers Who Bought This Also Bought