To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Mini plasmid kit
- Size:600 Preps
- Cat. no.:CA76539-236
Specifications
About this item
Isolate up to 25 µg plasmid DNA from 1 to 5 ml bacterial cultures using mini spin columns.
- Rapid - Purification of plasmid DNA in less than 30 minutes
- Safe - No Phenol/chloroform extractions
- Versatile - Spin and vacuum formats available
- High-quality - DNA is suitable for a variety of downstream applications
The E.Z.N.A.® Plasmid DNA Mini Kit I is designed to isolate up to 25 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in less than 30 minutes. Plasmid DNA purification follows the alkaline-lysis method and is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.