dNTP solutions, illustra™, Cytiva
Supplier: Cytiva
Certificates
About this item
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number28406501_HAND BOOK.pdf 28406502_HAND BOOK.pdf 28406522_HAND BOOK.pdf 28406532_HAND BOOK.pdf 28406541_HAND BOOK.pdf
Manual
CyDye Licensing Statements GE Healthcare Life Sciences Cytiva CyDye Licensing Statements Cytiva General Licensing Statements Licensing Statements GE Healthcare Life Sciences VWR GHS Compliance Letter (US)
Additional Product Details
GE Healthcare Next-Generation Sequencing (NGS) Solutions
Product Literature
Customers Who Bought This Also Bought