illustra™ PCR nucleotide mix dNTP set, Cytiva
Supplier: Cytiva
Certificates
About this item
PCR nucleotide mix dNTP set (2 mM, 10 mM, or 25 mM each A,C,G,T) for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready to use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberCA95038-268L_17211979
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought