About this item
This antibody has been tested to bind to monkeypox virus protein A35R (A33) dimeric protein (Catalog: S95L46).
Type: Primary
Antigen: Envelope A35R protein
Clonality: Monoclonal
Clone: RV10
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 Kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76729-602
- 76729-630
- Antigen name:
- Envelope A35R protein
- Envelope A35R protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- RV10
- RV10
- Host:
- Human
- Human
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Neutralisation:
- Yes
- Yes