To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:DYKDDDDK Tag
- Clonality:Monoclonal
- Clone:114F12C8
- Host:Rabbit
- Isotype:IgG
- Antigen symbol:DYKDDDDK Tag
- Conjugation:Biotin
- ELISA:Yes
- Size:40 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4 containing 1% BSA and 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:DYKDDDDK synthetic peptide coupled to KLH
- Purification:Protein A affinity column
- Cat. no.:76198-916
- Packaging:Vial
Specifications
About this item
GenScript MonoRab™ DYKDDDDK tag antibody [Biotin], mAb, rabbit specifically recognizes DYKDDDDK tags placed at C-terminal, N-terminal and internal regions of DYKDDDDK fusion proteins.
- Comprehensive pool
- against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: DYKDDDDK Tag
Clonality: Monoclonal
Clone: 114F12C8
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: