To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:His Tag
- Clonality:Monoclonal
- Clone:25B6E11
- Host:Rabbit
- Isotype:IgG
- Antigen symbol:His Tag
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:40 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4 containing 0.02% sodium azide
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:6x His synthetic peptide coupled to KLH
- Purification:Purified by Protein A affinity chromatograph
- Cat. no.:76197-310
- Packaging:Vial
Specifications
About this item
GenScript MonoRab™ anti-His tag (C-term) antibody (25B6E11), mAb, rabbit specifically recognizes His tags placed at C-terminal and internal regions of His-fusion
proteins. This product is not recommended for N-terminal His-fusion protein detection and nbsp.
- Comprehensive pool
- against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
The lyophilized product remains stable for up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 to 3 weeks at 2 to 8 °C or for up to 12 months at −20 °C or below. Upon receiving, spin the vial down to place the lyophilized antibody on the bottom of the vial. Reconstitute the antibody as mentioned above. Prepare single use aliquots (no less than 10 µl) in amber vials and freeze it immediately at −20 °C. The frozen aliquots should be thawed once with any remainder kept at 4 °C.
Type: Tag
Antigen: His Tag
Clonality: Monoclonal
Clone: 25B6E11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: