To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG
- Clonality:Monoclonal
- Clone:37C2
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit
- Antigen symbol:IgG
- Conjugation:iFluor® 488
- Size:200 µl
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (pH 7.2), containing 10 mg/ml BSA, 0.02% sodium azide and 50% glycerol
- Storage temperature:Store at −20 °C and it should be protected from prolonged exposure to light. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Concentration:0.5 mg/ml
- Shipping temperature:Ice
- Purification:Protein A affinity column
- Cat. no.:76510-072
- Packaging:Vial
Specifications
About this item
Rabbit IgG control is suitable for use as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, western blot, immunoprecipitation, immunodiffusion, and immunoelectrophoresis, flow cytometry and immunofluorescence. GenScript MonoRab™ rabbit IgG control [iFluor 488] (whole molecule), mAb is iFluor 488 conjugated MonoRab™ rabbit IgG control (whole molecule), mAb.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
.
Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone: 37C2
Conjugation: iFluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit