To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG
- Clonality:Monoclonal
- Clone:BP2a
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Unconjugated
- Size:1 mg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 2 years at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freeze and thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 1 mg/ml
- Shipping temperature:Ice
- Purification:Protein A affinity column
- Cat. no.:76510-022
- Packaging:Vial
Specifications
About this item
Mouse IgG2a control (whole molecule), purified is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, FACS, ICC immunoblotting, dot blot, western blot, immunoprecipitation, immunodiffusion, and immunoelectrophoresis. It may also be used as antigen or ligand in immunochemical conjugation reaction. GenScript mouse IgG2a control (whole molecule), purified is highly purified from cell culture in vitro under conditions free from animal-derived components through protein A affinity column.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
.
Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone: BP2a
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse