About this item
GenScript insulin antibody (6E9F1), mAb, mouse detects human insulin and slightly recognizes proinsulin, it has no cross-reactivity with C-peptide.
- High specificit
- High affinity
- High stability
GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including sandwich ELISA and western blot.
Type: Primary
Antigen: Insulin
Clonality: Monoclonal
Clone: 6E9F1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76509-842
- 76509-844
- Antigen name:
- Insulin
- Insulin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 6E9F1
- 6E9F1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa