To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bevacizumab
- Clonality:Monoclonal
- Clone:4H1
- Host:Mouse
- Isotype:IgG1
- Antigen symbol:Bevacizumab
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Bevacizumab
- Purification:Protein A affinity column
- Cat. no.:76509-976
- Packaging:Vial
Specifications
About this item
The product is specific for Bevacizumab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01976-40, Anti-Bevacizumab Antibody (6C3), mAb, Mouse.
- Anti-Idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
Antigen: Bevacizumab
Clonality: Monoclonal
Clone: 4H1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: