To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Camelid VHH
- Clonality:Monoclonal
- Clone:96A3F5
- Host:Rabbit
- Isotype:IgG
- Reactivity:Camel,Llama,Alpaca
- Antigen symbol:Camelid VHH
- Conjugation:iFluor® 647
- Flow cytometry:Yes
- Size:200 µl
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (pH 7.4), 10 mg/ml BSA, 0.02% sodium azide and 50% glycerol
- Storage temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Concentration:0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Llama VHH also called sdAb
- Purification:Protein A affinity column
- Cat. no.:76510-012
- Packaging:Vial
Specifications
About this item
The MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor 647], mAb is specific for Camelid VHH (Alpaca, Llama and Camel). This monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
- Anti-VHH Antibodies
- MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
- No cross reactivity with other species
- Specific to camelid IgG2 and IgG3
- Recognize conformational epitopes of the VHH domain
- Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.
Type: Primary
Antigen: Camelid VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: iFluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Alpaca;Llama;Camel