About this item
The product is specific for SARS-CoV-2 Nucleocapsid protein.
- COVID-19 antibodies
- Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
- Control antibodies and various antibody pairs available for ELISA-based detection
- Neutralizing Antibody Standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples
Genscript offers site-specific Neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type: Primary
Antigen: SARS-CoV-2 Nucleocapsid
Clonality: Monoclonal
Clone: 3F9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76510-122
- 76510-124
- Antigen symbol:
- SARS-CoV-2 Nucleocapsid
- SARS-CoV-2 Nucleocapsid
- Antigen name:
- SARS-CoV-2 Nucleocapsid
- SARS-CoV-2 Nucleocapsid
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 3F9
- 3F9
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa