To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:Beta Actin
- Clonality:Monoclonal
- Clone:79E2D7
- Host:Rabbit
- Isotype:IgG
- Antigen symbol:Beta Actin
- Conjugation:Unconjugated
- ELISA:Yes
- Size:40 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide coupled to KLH
- Purification:Protein A affinity column
- Cat. no.:76197-326
- Packaging:Vial
Specifications
About this item
GenScript MonoRab™ Beta-Actin Antibody, mAb, Rabbit reacts with mouse, human, hamster, and rat. It has not yet been tested in other species.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available.GenScript also offers a series of isotype control antibodies that can be used in your research.
Type: Control
Antigen: Beta Actin
Clonality: Monoclonal
Clone: 79E2D7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: