Sterilization Wraps, 2-Ply SMS, One-Step Heavy Duty
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificates
About this item
One-Step super duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
- Manufactured and packaged in a certified ISO 8 Cleanroom
- 2-ply
- Clean - no cellulose
- Strong, rip and tear resistant 88 gsm SMS material
- An extensive range of sizes available means less cutting
- Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
- Gram weight: 50 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Delivery: Certificate of Conformance sent with each shipment.
Packaging: Sub packed into sealed labeled poly bags with lot number and expiration date for complete traceability.
Customers Who Bought This Also Bought