Specifications
- Description:Total Chlorine test kit
- Method:colorimetric
- No. of Tests:50 avg.
Specifications
About this item
This chemical test kit that uses the DPD method to measure total chlorine. The test kit is supplied complete with all of the reagents and equipment necessary to perform approximately 50 tests.
- Pre-made reagents for ease of use
- Color comparison cube for simple determination
- Complete setup:All required materials are included with the test kit, such as the color comparison cube, and reagent dropper bottles
The chlorination of water supplies and polluted waters is used mainly to destroy or deactivate disease-producing microorganisms. Chlorine also serves to improve the quality of drinking water, as it reacts with ammonia, iron, manganese, sulfide, and some organic substances. Nevertheless, high amounts of chlorine will produce adverse effects like the formation of compounds that are potentially carcinogenic (e.g. chloroform) or harmful to aquatic life (e.g. chloramines). It remains essential to control the amount of added chlorine in order to fulfill the primary purpose of disinfecting while also minimizing any adverse effects.