ContactUsLinkComponent
Premium Comfort Cleanroom Socks
Premium Comfort Cleanroom Socks
Catalog #: 76795-312
Supplier:  Vidaro
CLEANROOM SOCKS REUSABLE POLYMIDE BLUE
Premium Comfort Cleanroom Socks
Catalog #: 76795-312
Supplier:  Vidaro
Specifications
  • Color:
    Blue
  • Description:
    Premium comfort cleanroom socks
  • Cat. no.:
    76795-312
Specifications
About this item
Premium, form-fitting polyamide cleanroom sock.
  • Heavy-duty polyamide cleanroom sock
  • Provides thickness and breathability for ultimate comfort
  • Flat toe seam for extra comfort
  • One size fits all
Premium Comfort cleanroom socks feature a 97% polyamide/3% elastine construction that provides quality form-fitting comfort and breathability for wearers.
Customers Who Bought This Also Bought