Multichannel Pipette Stand, Gilson
Catalog #: 76178-112
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Color:Transparent
- Description:Universal Multichannel Pipette Stand
- Cat. no.:76178-112
Specifications
About this item
The space-saving Universal Mulitchannel Stand accommodates any brand of 8- or 12-channel pipette, protecting them from damage and contamination.
- Design and materials allow the support edges to adapt to the lower part of virtually all pipettes
- Compact stand optimizes bench space and fits more easily into fume hoods
- It is made from rugged materials that are resistant to many chemicals
Because the stand holds a pipette vertically by its lower part, not the body as with most stands, the Universal Pipette Stand is very stable and ensures there is no risk to the cover and tip-holders.
Customers Who Bought This Also Bought