To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:15-LOX-2
- Description:Human 15-LOX-2 ELISA kit
- Size:96 tests
- Environmentally Preferable:
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human 15-LOX-2 ELISA kit exhibits high specificity and excellent specificity for the detection of human 15-LOX-2. No significant cross-reactivity or interference between 15-LOX-2 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0.313 - 20 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory status:RUO
- Cat. no.:76746-606
Human 15-LOX-2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human 15-LOX-2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.313 - 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human 15-LOX-2 ELISA kit (A74650) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human 15-LOX-2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for 15-LOX-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the 15-LOX-2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-15-LOX-2 Antibody, which binds the captured 15-LOX-2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of 15-LOX-2 captured in each well. The concentration of 15-LOX-2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.