Order Entry
ContactUsLinkComponent
Human Homocysteine ELISA Kit
Human Homocysteine ELISA Kit
Catalog #: 76740-376
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
ELISA KIT HUMAN HCY
Human Homocysteine ELISA Kit
Catalog #: 76740-376
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Reactivity:
    Human
  • Sensitivity:
    4.688 pmol/ml
  • Target protein:
    Homocysteine
  • Description:
    Human homocysteine ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Homocysteine ELISA Kit exhibits high specificity and excellent specificity for the detection of human Homocysteine. No significant cross-reactivity or interference between Homocysteine and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    7.813 - 500 pmol/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Regulatory status:
    RUO
  • Cat. no.:
    76740-376

Specifications

About this item

Human Homocysteine ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Homocysteine in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 7.813 to 500 pmol/ml
  • Sensitivity: 4.688 pmol/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Homocysteine ELISA kit (A79813) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Homocysteine in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Homocysteine antigen. During the incubation, Homocysteine present in the samples or standards competes with the fixed amount of immobilized Homocysteine for binding sites on the Biotinylated Anti-Homocysteine Antibody. The more Homocysteine present in a sample or standard, the less Biotinylated Anti-Homocysteine Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Homocysteine Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Homocysteine present in each sample or standard. The concentration of Homocysteine can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Customers Who Bought This Also Bought