Human Acetylated p53 ELISA Kit
Catalog #: 76730-676
Supplier: ANTIBODIES.COM LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Reactivity:Human
- Sensitivity:28.125 pg/ml
- Target protein:Acetylated p53
- Description:Human acetylated p53 ELISA kit
- Format:Pre-coated
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human Acetylated p53 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Acetylated p53. No significant cross-reactivity or interference between Acetylated p53 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Type:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:46.875 - 3000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Regulatory status:RUO
Specifications
About this item
This Human Acetylated p53 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Human Acetylated p53 in plasma, tissue homogenates and other biological fluids.
Customers Who Bought This Also Bought