Mouse IgG ELISA Kit
Catalog #: 76737-192
Supplier: ANTIBODIES.COM LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Sensitivity:0.938 ng/ml
- Target protein:IgG
- Description:Mouse IgG ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids.
- Detection method:Colorimetric microplate reader
- Time to Results:4 h 30 min
- Shelf life:6 months (4 °C)
- Detection range:1.563 - 100 ng/ml
- Storage temperature:Store at 4 °C. Do not use past expiration date
- Regulatory status:RUO
- Cat. no.:76737-192
Specifications
About this item
This Mouse IgG ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse IgG in plasma, tissue homogenates and other biological fluids.
Customers Who Bought This Also Bought