Order Entry
ContactUsLinkComponent
Anti-SARS-CoV-2 (N) Immunoglobulin ELISA Kit
Anti-SARS-CoV-2 (N) Immunoglobulin ELISA Kit
Catalog #: 76735-452
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Anti-SARS-CoV-2 (N) Immunoglobulin ELISA Kit
Catalog #: 76735-452
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Reactivity:
    Other
  • Target protein:
    Anti-SARS-CoV-2 (N) Immunoglobulin
  • Description:
    Anti-SARS-CoV-2 (N) immunoglobulin ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Anti-SARS-CoV-2 (N) Immunoglobulin ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Anti-SARS-CoV-2 (N) Immunoglobulin. No significant cross-reactivity or interference between Anti-SARS-CoV-2 (N) Immunoglobulin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Regulatory status:
    RUO
  • Cat. no.:
    76735-452

Specifications

About this item

Anti-SARS-CoV-2 (N) Immunoglobulin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of Anti-SARS-CoV-2 (N) Immunoglobulin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Customers Who Bought This Also Bought