Order Entry
ContactUsLinkComponent
Mouse beta Crosslaps/beta CTx ELISA Kit
Mouse beta Crosslaps/beta CTx ELISA Kit
Catalog #: 76732-602
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Mouse beta Crosslaps/beta CTx ELISA Kit
Catalog #: 76732-602
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Reactivity:
    Mouse
  • Sensitivity:
    46.875 pg/ml
  • Target protein:
    beta Crosslaps/beta CTx
  • Description:
    Mouse beta crosslaps/beta CTx ELISA kit
  • Format:
    Pre-coated
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse beta Crosslaps/beta CTx ELISA kit exhibits high specificity and excellent specificity for the detection of mouse beta Crosslaps/beta CTx. No significant cross-reactivity or interference between beta Crosslaps/beta CTx and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Type:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    78.125 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Regulatory status:
    RUO

Specifications

About this item

Mouse beta Crosslaps/beta CTx ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse beta Crosslaps/beta CTx in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 78.125 - 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse beta Crosslaps/beta CTx ELISA kit (A75978) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse beta Crosslaps/beta CTx in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with beta Crosslaps/beta CTx antigen. During the incubation, beta Crosslaps/beta CTx present in the samples or standards competes with the fixed amount of immobilized beta Crosslaps/beta CTx for binding sites on the Biotinylated Anti-beta Crosslaps/beta CTx Antibody. The more beta Crosslaps/beta CTx present in a sample or standard, the less Biotinylated Anti-beta Crosslaps/beta CTx Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-beta Crosslaps/beta CTx Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of beta Crosslaps/beta CTx present in each sample or standard. The concentration of beta Crosslaps/beta CTx can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Customers Who Bought This Also Bought