Order Entry
ContactUsLinkComponent
Mouse Anti-KLH IgG ELISA Kit
Mouse Anti-KLH IgG ELISA Kit
Catalog #: 76733-764
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Mouse Anti-KLH IgG ELISA Kit
Catalog #: 76733-764
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Reactivity:
    Mouse
  • Sensitivity:
    0.938 ng/ml
  • Target protein:
    Anti-KLH IgG
  • Description:
    Mouse anti-KLH IgG ELISA kit
  • Format:
    Pre-coated
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Type:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1.563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Anti-KLH IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-KLH IgG in serum, plasma, tissue homogenates, and other biological fluids.

Customers Who Bought This Also Bought