Order Entry
ContactUsLinkComponent
Human Cysteine Dioxygenase CDO1 ELISA Kit
Human Cysteine Dioxygenase CDO1 ELISA Kit
Catalog #: 76739-600
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
ELISA KIT HUMAN CDO1
Human Cysteine Dioxygenase CDO1 ELISA Kit
Catalog #: 76739-600
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Reactivity:
    Human
  • Sensitivity:
    0.191 ng/ml
  • Target protein:
    Cysteine Dioxygenase CDO1
  • Description:
    Human cysteine dioxygenase CDO1 ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Cysteine Dioxygenase CDO1 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Cysteine Dioxygenase CDO1. No significant cross-reactivity or interference between Cysteine Dioxygenase CDO1 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0.5 - 32 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Regulatory status:
    RUO
  • Cat. no.:
    76739-600

Specifications

About this item

Human Cysteine Dioxygenase CDO1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cysteine Dioxygenase CDO1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.5 to 32 ng/ml
  • Sensitivity: 0.191 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Cysteine Dioxygenase CDO1 ELISA kit (A77864) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cysteine Dioxygenase CDO1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cysteine Dioxygenase CDO1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cysteine Dioxygenase CDO1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cysteine Dioxygenase CDO1 Antibody, which binds the captured Cysteine Dioxygenase CDO1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cysteine Dioxygenase CDO1 captured in each well. The concentration of Cysteine Dioxygenase CDO1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Customers Who Bought This Also Bought