Eisco® Small Anemometer
Catalog #: 470343-686
Supplier: Wards
Specifications
- Description:Small anemometer
- Height:178 mm (7")
- Cat. no.:470343-686
- Diameter:216 mm (8.5")
Specifications
About this item
Measures wind speed and is portable for outdoor use.
- Perfect for students of any age
- Corrosion resistant materials for prolonged outdoor use
This small anemometer is perfect for younger students to use in determing wind speed. Cups are mounted on horizontal arms extending from the axis with one red cup being used to determine the number of rotations.
Customers Who Bought This Also Bought