To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:MAGEA12
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Environmentally Preferable:
- Epitope:121-210/314
- Form:Liquid
- Gene ID:4111
- Antigen synonyms:MAGE12F antigen|MAGE 12 antigen|Melanoma associated antigen 12|MAGEA12|Melanoma-associated antigen 12|MAGE-12 antigen|CT1.12|Cancer/testis antigen 1.12|MAGE12|MAGAC_HUMAN|Cancer/testis antigen family 1 member 12|MAGE12F|Melanoma antigen family A 12|MAGE12 antigen
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular weight:35 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MAGEA12
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76118-796
- Packaging:Vial
Not known, though may play a role in tumor transformation or aspects of tumor progression.Tissue specificity: Expressed in many tumors of several types, such as melanoma, head and neck squamous cell carcinoma, lung carcinoma and breast carcinoma, but not in normal tissues except for testes.
Type: Primary
Antigen: MAGEA12
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-210/314
Host: Rabbit
Isotype: IgG
Reactivity: Human