To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- W×L:11.75×12.13"
- Color:Red
- Description:Bio-wipe bag
- Cat. no.:89092-524
- Supplier No.:MLBM015111
Specifications
About this item
Quick, economical cleanup for biohazardous spills.
- All-in-one absorbent wipe pad and heavy-duty bag
- Absorbent wipe pad holds 1/2 pint (8oz. or 1 cup) liquid capacity
- Bag features adhesive seal to ensure total containment
- Instructions clearly printed on bag
- Integral ambidextrous mitt inside bag
- Makes compliance with OSHA Bloodborne Pathogens Standard easy for small spills
- Do not use for containment of needles or other sharps
Absorbent pad (11.75×9") capacity is approximately 1/2 pint. An adhesive strip provides secure closure for safe containment and proper disposal, and an ambidextrous mitt is included inside the bag for better maneuverability. Made in the USA.