Specifications
- Description:Kitronik smart greenhouse
- Cat. no.:470338-524
Specifications
About this item
Build automated growing systems that react to changeable environmental condition.
- Compatible with both micro:bit V1 and micro:bit V2
- Comes with detailed assembly and coding instructions
- Seven online MakeCode tutorials
The Kitronik smart greenhouse kit for the BBC micro:bit provides an exciting way to learn about the relationship between plants and their environment. The user will learn how to monitor environmental conditions and then how to build automated systems that react to them. Grow a crop of plants with this smart greenhouse that can be coded to monitor and automatically water them so that you can get them growing perfectly. This kit is compatible with both the micro:bit V1 and the new micro:bit V2. The kit comprises a two-part plastic greenhouse, Kitronik environmental control board, water pump, Kitronik ZIP Stick, and Mini Prong soils moisture sensor. Also included are 5 crocodile clips, ZIP extension cable and a small screwdriver.