Tempo iOligo™ Human iPSC derived Oligodendrocyte Progenitor Cells (OPCs)
Catalog #: 76683-018
Supplier: TEMPO BIOSCIENCE, INC
CAS Number:
Specifications
- Description:Human iPSC-derived oligodendrocyte progenitor cells (OPCs)
- Cat. no.:76683-018
Specifications
About this item
Human iPSC-derived Oligodendrocyte Progenitor Cells (OPCs).
- Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought