Tempo iMono™ Human iPSC derived CD14+ Monocytes
Catalog #: 76683-004
Supplier: TEMPO BIOSCIENCE, INC
CAS Number:
Specifications
- Description:Human iPSC-derived CD14+ monocytes
Specifications
About this item
Human iPSC-derived CD14+ Monocytes.
- Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought