TempoCal™ Rapid Calcium Biosensor Assay using Immortalized Human Neuroepithelial Cells
Catalog #: 76682-980
Supplier: TEMPO BIOSCIENCE, INC
CAS Number:
Specifications
- Description:TempoCal biosensor transiently expressed in TempoRapid neuroepithelial cells
Specifications
About this item
TempoCal biosensor transiently expressed in TempoRapid neuroepithelial cells
- 1 Cryovial = ~1.025×10^⁶
- cells/ml
Product Type: human cells
Long-Term Storage: liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought