Specifications
- Description:AimStrip™ Xylazine Urine Dipstick
- Size:25/Kit
- Cat. no.:76717-408
Specifications
About this item
AimStrip® Xylazine Urine/Powder Dipstick, 25/Kit. Cut-off concentration of 1000 ng/ml.
- XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.
- Human intoxication with XYL has been reported sporadically over the past several decades
- Noted in literature describing drug overdose deaths in Philadelphia as early as 2006
- Since mid-2010's, XYL has been noticed by people who inject drugs (PWID) and public health practitioners as an increasingly commonplace additive in the street opioid supply of Philadelphia.
- Recent reports from Connecticut implicated XYL in a rising fraction of overdose deaths in 2019–2020
- In Puerto Rico, heroin is commonly adulterated with XYL
- Frequently found in speedball (a cocaine and heroin mixture)
A rapid test for the qualitative detection of xylazine in human urine and powder. For forensic use only, not for in vitro diagnostic use. A rapid chromatographic immunoassay for the detection of xylazine in human urine and powders at a cut-off concentration of 1000 ng/ml. The Xylazine (XYL) Rapid Test Dipstick is a rapid screening test that can be performed without the use of an instrument. The test utilizes the antibody to selectively detect elevated levels of xylazine in urine and powders.
XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.