Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Epidermal Growth Factor (from E. coli)
Human Recombinant Epidermal Growth Factor (from E. coli)
Catalog # CACB354052
Supplier:  Corning
CAS Number:  
Human Recombinant Epidermal Growth Factor (from E. coli)
Catalog # CACB354052
Supplier:  Corning
Supplier Number:  354052
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein synonyms:
    Epidermal Growth Factor
  • Protein/peptide name:
    EGF
  • Purity:
    Purity > 98% by SDS-PAGE
  • Cat. no.:
    CACB354052
  • Supplier No.:
    354052

Specifications

About this item

Epidermal growth factor (EGF) is a low-molecular weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

  • Media additive

EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

Mouse natural EGF are tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. Human recombinant EGF are shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.