About this item
HyClone™ standard cell culture grade FBS is filtered through three sequential 100 µm (0,1 µm) pore-size rated filters.
- Sourced in the USA with complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
HyClone Standard Fetal Bovine Serum (FBS), U.S. origin is an economical alternative to characterized and defined FBS. Standard FBS is ideal for research applications.
Standard FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC) and IgG. Included assays are subject to change without notice.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76324-888
- CA82013-574
- CA76324-890
- CA16777-532
- CA16777-534
- CA76236-308
- Origin:
- US
- US
- US
- US
- US
- US
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Less than –10 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
Specifications
Related Information
Cytiva BioXtra and Pall Life Sciences Products CA Cytiva BioXtra and Pall Life Sciences Products US
Promotion Material
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Cytiva HyClone Media and Supplements HyClone™ Cell Culture Serum products